Learn More
Electron Microscopy Sciences PCR 100-Well Storage Rack, Orange
SDP
Catalog No. 501899015
Encompass_Preferred
Polypropylene rack features durable three point hinges and clasp for secure lid closure. Holds 1.5 to 2.0 ml microtubes and cryovials. User friendly rack features molded gridlines on the lid, imprinted coordinates on the bottom of base, and imprinted and raised coordinates on the box. Rack measures 5.625 x 5.875 x 2.25".
Color: Orange
Catalog No. 50-189-9015 Supplier Electron Microscopy Sciences Supplier No. 61601O
Polypropylene rack features durable three point hinges and clasp for secure lid closure. Holds 1.5 to 2.0 ml microtubes and cryovials. User friendly rack features molded gridlines on the lid, imprinted coordinates on the bottom of base, and imprinted and raised coordinates on the box. Rack measures 5.625 x 5.875 x 2.25".
Color: Orange