Learn More
Sigma Aldrich Fine Chemicals Biosciences PCR 50 - 2000 bp Marker fo
Catalog No. 502904376
Encompass
Sigmas PCR 50 - 2 000 bp Marker contains 8 dsDNA fragments from 50 - 2 000 bases evenly spaced. five mul of the ladder can be loaded directly in a single lane on an agarose or polyacrylamide gel.
Catalog No. 50-290-4376 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P95771VL
Sigmas PCR 50 - 2 000 bp Marker contains 8 dsDNA fragments from 50 - 2 000 bases evenly spaced. five mul of the ladder can be loaded directly in a single lane on an agarose or polyacrylamide gel.