Learn More
Integra Biosciences Corp 96 Well PCR Cooling Plates
SDP
Supplier:  Integra Biosciences Corp 6250
Encompass
Compatible with VIAFLO 96/384 and ASSIST PLUS. PCR aluminum cooling block to keep reagents in various PCR plates chill. Compatible with 0.2 ml PCR tubes or 96 well PCR plates. Support for non-skirted 96 well PCR plates.
Catalog No. NC1425859