Learn More
Sigma Aldrich Fine Chemicals Biosciences PCR 96 Well rack assorted
Catalog No. 502900257
Encompass
Sturdy rack for working with 0.2 mL tubes. Accommodates the standard 8 or 12 tube strips. Perfect for pre- or post-PCR sampling applications. The tube wells are easily identified with imprinted numbers and letters. The rack features a removable hinged lid.
Catalog No. 50-290-0257 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R69015EA
Sturdy rack for working with 0.2 mL tubes. Accommodates the standard 8 or 12 tube strips. Perfect for pre- or post-PCR sampling applications. The tube wells are easily identified with imprinted numbers and letters. The rack features a removable hinged lid.