Learn More
Air Science Usa Cart-36 Mobile Base Stand, Durable Powder Coated Steel, Open Frame Design, 36 in
SDP
Catalog No. NC2133431
Encompass_Preferred
The Cart-36 Mobile Base Stand provides a sturdy and ergonomic foundation for 36-inch wide Purair PCR Laminar Flow Cabinets and other compatible lab enclosures. Its robust open-frame design is made of durable powder-coated steel, featuring a lower shelf for extra storage. With mobility options like locking casters, this stand offers flexibility and stability, ensuring secure placement while enabling easy repositioning within the laboratory
This item is not returnable. View return policy
Catalog No. NC2133431 Supplier Air Science Usa Supplier No. CART36
The Cart-36 Mobile Base Stand provides a sturdy and ergonomic foundation for 36-inch wide Purair PCR Laminar Flow Cabinets and other compatible lab enclosures. Its robust open-frame design is made of durable powder-coated steel, featuring a lower shelf for extra storage. With mobility options like locking casters, this stand offers flexibility and stability, ensuring secure placement while enabling easy repositioning within the laboratory