Learn More
Teknova PCR Certified Water. 50mL Tubes, 25-Pack, DNase/RNase Tested, Sterile.
SDP
Catalog No. NC3050430
Encompass
PCR testing is an essential part of quality control for all water products, and the only way to guarantee that a product is safe to use for PCR-based applications. When you choose PCR Certified products, you can be confident that they have undergone a rigorous testing process to ensure that they are free of contamination.
Catalog No. NC3050430 Supplier Teknova Supplier No. 2W3336
PCR testing is an essential part of quality control for all water products, and the only way to guarantee that a product is safe to use for PCR-based applications. When you choose PCR Certified products, you can be confident that they have undergone a rigorous testing process to ensure that they are free of contamination.