Learn More
Thomas Scientific PCR Clean Spray, 4X500 mL Refill
SDP
Catalog No. NC1633138
Encompass
PCR Clean is ready-to-use solution for eliminating DNA from any surface. DNA is removed within seconds after use. The solution contains a surfactant and a non-alkaline and non-carcinogenic agent. PCR Clean is intended for use at PCR workstations, lab devices, pipettors, etc.
Catalog No. NC1633138 Supplier Thomas Scientific Supplier No. C791Q55
PCR Clean is ready-to-use solution for eliminating DNA from any surface. DNA is removed within seconds after use. The solution contains a surfactant and a non-alkaline and non-carcinogenic agent. PCR Clean is intended for use at PCR workstations, lab devices, pipettors, etc.