Learn More
Thomas Scientific PCR CleanT Spray, 250mL
SDP
Catalog No. NC1590308
Encompass
Contaminant DNA is especially problematic in the highly sensitive PCR technique, as its removal has proven to be no trivial matter. Originating from aerosolized fragments, contaminant DNA can lead to cross contamination between samples thus resulting in inaccurate data or PCR artefacts. Even a single DNA molecule can be detected in the amplification process leading to widespread problems throughout the testing procedure. PCR Clean™ is ready-to-use solution for eliminating DNA from any surface. DNA is removed within seconds after use. The solution contains a surfactant and a non-alkaline and non-carcinogenic agent. PCR Clean™ is intended for use at PCR workstations, lab devices, pipettors, etc. Benefits Highly effective PCR Clean™ is effective against amplicon, plasmid, or genomic DNA and RNA from most surfaces with the exception of light or non-ferrous metals. Convenient and ready-to-use PCR Clean™ is ready-to-use for eliminating DNA and RNA from suitable surfac
Catalog No. NC1590308 Supplier Thomas Scientific Supplier No. C791Q54
Contaminant DNA is especially problematic in the highly sensitive PCR technique, as its removal has proven to be no trivial matter. Originating from aerosolized fragments, contaminant DNA can lead to cross contamination between samples thus resulting in inaccurate data or PCR artefacts. Even a single DNA molecule can be detected in the amplification process leading to widespread problems throughout the testing procedure. PCR Clean™ is ready-to-use solution for eliminating DNA from any surface. DNA is removed within seconds after use. The solution contains a surfactant and a non-alkaline and non-carcinogenic agent. PCR Clean™ is intended for use at PCR workstations, lab devices, pipettors, etc. Benefits Highly effective PCR Clean™ is effective against amplicon, plasmid, or genomic DNA and RNA from most surfaces with the exception of light or non-ferrous metals. Convenient and ready-to-use PCR Clean™ is ready-to-use for eliminating DNA and RNA from suitable surfac