Learn More
Thomas Scientific PCR CleanT Wipes, 5 x 120 refill sachets
SDP
Catalog No. NC1590311
Encompass
The presence of contaminant DNA at a PCR work area can result in false positive results and misinterpretation. DNA contaminations are usually sustainable and their removal has proven to be no trivial matter. PCR Clean™ Wipes are ready-to-use for eliminating DNA from surfaces like glass, ceramic, plastic, rubber, steel and precious metal. DNA and RNA are removed within seconds after use, and enzymes like DNases and RNases are inactivated. PCR Clean™ Wipes are the simple solution for cleaning and decontaminating surfaces in a PCR laboratory. There’s no need for multiple products when you have the convenience of a simple-to-use wipe. When you use a sponge or even paper towel, you can spread the DNA contamination from one surface to another potentially increasing the contaminated surface area. Recommended Use / Scope Applicable in research and industry only. Not recommended for clinical applications. Use as directed. Always test a small area first for stability and colora
This item is not returnable. View return policy
Catalog No. NC1590311 Supplier Thomas Scientific Supplier No. C791Q58
The presence of contaminant DNA at a PCR work area can result in false positive results and misinterpretation. DNA contaminations are usually sustainable and their removal has proven to be no trivial matter. PCR Clean™ Wipes are ready-to-use for eliminating DNA from surfaces like glass, ceramic, plastic, rubber, steel and precious metal. DNA and RNA are removed within seconds after use, and enzymes like DNases and RNases are inactivated. PCR Clean™ Wipes are the simple solution for cleaning and decontaminating surfaces in a PCR laboratory. There’s no need for multiple products when you have the convenience of a simple-to-use wipe. When you use a sponge or even paper towel, you can spread the DNA contamination from one surface to another potentially increasing the contaminated surface area. Recommended Use / Scope Applicable in research and industry only. Not recommended for clinical applications. Use as directed. Always test a small area first for stability and colora