Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ PCR Purification Wash Buffer W1

Catalog No. K310006
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K310006 40 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K310006 Supplier Invitrogen™ Supplier No. K310006
Only null left
Add to Cart
Add to Cart

The buffer is activated by the addition of 160 mL of 100% ethanol.

The buffer is activated by the addition of 160 mL of 100% ethanol.

Not High Throughput-Compatible (Manual)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Wash Buffer
Content And Storage Store the wash buffer at room temperature
Sample Type PCR Products
Final Product Type DNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line PureLink
Product Type PCR Purification Wash Buffer W1
Quantity 40 mL
Shipping Condition Room Temperature
System Type PureLink™

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.