Learn More
Research Products International Corp UltraFlux Rigid PCR Clear Tube Strips with Clear Caps, 120 Strips of 8 Tubes per Package
SDP
Catalog No. 50998905
Encompass
UltraFlux Rigid PCR Tube Strips are easier to work with than strips designed to snap apart. These eight thin wall tubes are joined together by a strong double link that reduces flexibility and provides more convenient handling.
Catalog No. 50-998-905 Supplier Research Products International Corp Supplier No. 249950
UltraFlux Rigid PCR Tube Strips are easier to work with than strips designed to snap apart. These eight thin wall tubes are joined together by a strong double link that reduces flexibility and provides more convenient handling.