Learn More
Research Products International Corp PCR Combi-Rack, 5 per Case
SDP
Catalog No. 50998911
Encompass
PCR Combi-Rack is ideal for carrying, storing, freezing and transporting sample tubes. Polypropylene rack holds up to 96 x 0.2ml tubes in 8 x 12 format. Withstands temperatures from -80°C to +120°C and is autoclavable. Features alpha numeric indexing for quick identification. Supplied with PVC base and cover for storage in a refrigerator or freezer at temperatures to -30°C. Base can also be filled with ice for use as a benchtop workstation. Accepts all 96 or 384 well plates.
Catalog No. 50-998-911 Supplier Research Products International Corp Supplier No. 353080
PCR Combi-Rack is ideal for carrying, storing, freezing and transporting sample tubes. Polypropylene rack holds up to 96 x 0.2ml tubes in 8 x 12 format. Withstands temperatures from -80°C to +120°C and is autoclavable. Features alpha numeric indexing for quick identification. Supplied with PVC base and cover for storage in a refrigerator or freezer at temperatures to -30°C. Base can also be filled with ice for use as a benchtop workstation. Accepts all 96 or 384 well plates.