Learn More
MINERVA BIOLABS INC PCR Cycler Check™ Advance 6 strips, 8 vials each
Catalog No. NC2606961
Encompass_Preferred
Validation tool for conventional PCR cyclers according to legal requirements (e.g. ISO 17025, EN 45001, ISO 13485, ISO/TS 20836:2007, GLP, GMP, IQ/OQ/PQ, and others).Kit ComponentsLyophilized primer sets, polymerase and nucleotidesRehydration bufferSize marker for optimized gel evaluation
Catalog No. NC2606961 Supplier MINERVA BIOLABS INC Supplier No. 572102
Validation tool for conventional PCR cyclers according to legal requirements (e.g. ISO 17025, EN 45001, ISO 13485, ISO/TS 20836:2007, GLP, GMP, IQ/OQ/PQ, and others).Kit ComponentsLyophilized primer sets, polymerase and nucleotidesRehydration bufferSize marker for optimized gel evaluation