Learn More
Enzo Life Sciences PCR decontamination kit (100 Reactions)
SDP
Catalog No. 502013832
Encompass_Preferred
The PCR decontamination kit uses a dsDNase to remove contaminating DNA from mastermixes. The double-strand specific property allows decontamination in the presence of primers and probe.The dsDNase is irreversibly inactivated by heating to 60°C in the presence of DTT, ensuring that DNA template added to the mastermix after inactivation will not be digested. The mastermix can be used immediately after decontamination to run PCR reactions. AMPIGENE Taq polymerases have been treated to remove host DNA and additional clean-up is not necessary. The PCR decontamination kit is recommended for decontamination of primers or competitor’s PCR mixes.
Catalog No. 50-201-3832 Supplier Enzo Life Sciences Supplier No. ENZKIT1370100
The PCR decontamination kit uses a dsDNase to remove contaminating DNA from mastermixes. The double-strand specific property allows decontamination in the presence of primers and probe.The dsDNase is irreversibly inactivated by heating to 60°C in the presence of DTT, ensuring that DNA template added to the mastermix after inactivation will not be digested. The mastermix can be used immediately after decontamination to run PCR reactions. AMPIGENE Taq polymerases have been treated to remove host DNA and additional clean-up is not necessary. The PCR decontamination kit is recommended for decontamination of primers or competitor’s PCR mixes.