Learn More
IBI SCIENTIFIC 3P Gel Extraction & PCR Cleanup Kit, 300 preps
Catalog No. NC0326720
Encompass_Preferred
DNA Purification Kit, PCR Clean-Up/Gel Extraction, IBI Scientific, Sample Size: 300mg agarose gel or 100ul PCR product, Binding Capacity: 10ug, Operation Time: 20min, Expectant Yield: 80-90% for gel extraction and 90-95% for PCR clean-up.
Catalog No. NC0326720 Supplier IBI SCIENTIFIC 3P Supplier No. IB47030
DNA Purification Kit, PCR Clean-Up/Gel Extraction, IBI Scientific, Sample Size: 300mg agarose gel or 100ul PCR product, Binding Capacity: 10ug, Operation Time: 20min, Expectant Yield: 80-90% for gel extraction and 90-95% for PCR clean-up.