Learn More
IBI SCIENTIFIC 3P Gel Extraction & PCR Cleanup Kit, 100 preps
Catalog No. NC0527389
Encompass_Preferred
Gel/PCR DNA Fragment Extraction kit 100 preps Sample: Up to 300mg agarose gel slice Up to 100ul PCR product or other enzymatic reaction Format: Spin Column Operation: Centrifuge/Vacuum manifold Binding
Catalog No. NC0527389 Supplier IBI SCIENTIFIC 3P Supplier No. IB47020
Gel/PCR DNA Fragment Extraction kit 100 preps Sample: Up to 300mg agarose gel slice Up to 100ul PCR product or other enzymatic reaction Format: Spin Column Operation: Centrifuge/Vacuum manifold Binding