Learn More
Bioworld PCR DNA PURIFICATION KIT
Catalog No. 50196949
Encompass_Preferred
For the rapid purification of amplified DNA products from PCR mixes Purify amplified DNA ranging from 100 bp -15,000bp in size; Fast and efficient spin column format. FEATURES AND BENEFITS Fast and easy processing - Rapid spin-column fo...
Catalog No. 50-196-949 Supplier Bioworld Supplier No. 107600921
For the rapid purification of amplified DNA products from PCR mixes Purify amplified DNA ranging from 100 bp -15,000bp in size; Fast and efficient spin column format. FEATURES AND BENEFITS Fast and easy processing - Rapid spin-column fo...