Learn More
LGC GENOMICS LLC (Lucigen) MasterAmp 10X PCR Enhancer, 10 mL(with betaine) improves the specificity and yield of PCR amplification reactions, especially templates containing a high GC content or secondary structure.
Catalog No. NC9217536
Encompass
For use with the MasterAmp™ PCR Optimization Kits.
This item is not returnable. View return policy
Catalog No. NC9217536 Supplier LGC GENOMICS LLC Supplier No. ME81210
For use with the MasterAmp™ PCR Optimization Kits.