Learn More
Bellco Glass, Inc. PCR film scraper
SDP
Catalog No. 502329386
Encompass_Preferred
PCR film scraper. Using with sealing film to make the sealing film fit better with the PCR plate. Made of TPE.
Catalog No. 50-232-9386 Supplier Bellco Glass, Inc. Supplier No. 451400471
PCR film scraper. Using with sealing film to make the sealing film fit better with the PCR plate. Made of TPE.