Learn More
Chemglass Life Sciences PCR Film Scraper/Application Tool, 70 x 78mm
SDP
Catalog No. 501897748
Encompass_Preferred
PCR Film scraper tool aides in the application of sealing film on PCR plates. Tool is made from Blue TPE.
Catalog No. 50-189-7748 Supplier Chemglass Life Sciences Supplier No. CGN400100P
PCR Film scraper tool aides in the application of sealing film on PCR plates. Tool is made from Blue TPE.