Learn More
Enzo Life Sciences PCR & Gel clean-up kit (50tests)
SDP
Catalog No. 502013733
Encompass_Preferred
PCR & Gel clean-up columns use silica membrane technology in a mini spin column format to deliver easy and efficient DNA purification from PCR products, agarose gels,  and labeling reactions. Sample material: <400 µl PCR reaction mixture, <400mg agarose gel Fragment size: 50bp - ~20kbp Recovery: 70 - 95% Elution volume: 15 - 50 µl Preparation time: 10 min for 6 preps Binding capacity: 25 µg PCR & Gel clean-up columns have been validated for use with our CGH Labeling Kit, Prod. No ENZ-42671. Assay Time: <10 min.
Catalog No. 50-201-3733 Supplier Enzo Life Sciences Supplier No. ENZGEN1000050
PCR & Gel clean-up columns use silica membrane technology in a mini spin column format to deliver easy and efficient DNA purification from PCR products, agarose gels,  and labeling reactions. Sample material: <400 µl PCR reaction mixture, <400mg agarose gel Fragment size: 50bp - ~20kbp Recovery: 70 - 95% Elution volume: 15 - 50 µl Preparation time: 10 min for 6 preps Binding capacity: 25 µg PCR & Gel clean-up columns have been validated for use with our CGH Labeling Kit, Prod. No ENZ-42671. Assay Time: <10 min.