Learn More
Growcells PCR GRADE H2O PK100
Catalog No. 501034866
Encompass_Preferred
Sterile PCR grade nuclease free water. Prepared with 18.2 megOhm, 0.1 micron sterile filtered water and autoclaved.
Catalog No. 50-103-4866 Supplier Growcells Supplier No. PCPW1000
Sterile PCR grade nuclease free water. Prepared with 18.2 megOhm, 0.1 micron sterile filtered water and autoclaved.