Learn More
Poltex PCR Hood Organizer
SDP
Catalog No. 22000221
Change view
Click to view available options
PCR Hood Organizer
Catalog No.
22-000-221 PCR Hood Organizer
1 options
Catalog No. 22-000-221 Supplier Poltex Supplier No. PCRHOOD
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
To organize pipettors, tips and tubes inside PCR hood. Poltex PCR Hood Organizer is used to store items on flat top and underneath the unit.
  • Made from durable, break-resistant, and alcohol-resistant 0.25 in. thick PETG
  • Holds 5 pipettors at an ergonomic angle toward user as well as up to 4 multi-channel pipettors off the sides
  • Diameter (Pipettor holes): 1 in.
Specifications
Product Type In-Hood Organizer, Pipette Storage
Dimensions (L x W x H) 7.5 x 12 x 11.75 in.
Holds 5 pipettors, 4 multi-channel pipettors, tip boxes, tube racks
Material PETG