Learn More
Viagen Biotech Inc DirectPCR Lysis Reagents
SDP
Catalog No. NC0452067
Encompass
DirectPCR Lysis Reagents are cell lysis reagents designed for direct PCR amplification from a variety of sample types including blood, bacteria, yeast, tissue, plants, and insects. The reagents lyse cells and degrade proteins while protecting nucleic acids from degradation. They allow PCR amplification directly from crude samples without DNA extraction or purification. The reagents are available in several formulations optimized for different sample types. They provide a fast and simple method for PCR amplification from samples containing proteins, lipids, polysaccharides, or other PCR inhibitors.
Catalog No. NC0452067 Supplier Viagen Biotech Inc Supplier No. 301C
DirectPCR Lysis Reagents are cell lysis reagents designed for direct PCR amplification from a variety of sample types including blood, bacteria, yeast, tissue, plants, and insects. The reagents lyse cells and degrade proteins while protecting nucleic acids from degradation. They allow PCR amplification directly from crude samples without DNA extraction or purification. The reagents are available in several formulations optimized for different sample types. They provide a fast and simple method for PCR amplification from samples containing proteins, lipids, polysaccharides, or other PCR inhibitors.