Learn More
Viagen Biotech Inc PCR LYSIS RGNT(MOUSE EAR
SDP
Supplier:  Viagen Biotech Inc 402E
Encompass
DirectPCR Lysis Reagent (Ear) 50 ml. Viagen DirectPCRa DNa Extraction System is a single-tube system for rapid preparation of DNa from mouse tails, ear pieces, yolk sac, and culture cells. The patent-pending components developed by scientists at Viagen Biotech Inc. allow the resulting DNa extracts to be compatible with genomic PCR for genotyping. Crude extracts of biological samples are not compatible with many molecular biology-grade reactions such as polymerase chain reaction (PCR), in part due to inhibitors contained in crude extracts. The DirectPCR reagents not only mediate the rapid lysis of biological samples but also contain inhibitors that effectively suppress the inhibitory activities of crude lysates for PCR amplification, while maximally maintaining the integrity of released genomic DNa. The patent-pending simple procedure completely eliminate any solution transfer or tube-opening steps, providing you with substancial extra time.
Catalog No. NC9771658