Learn More
New England Biolabs, Inc. PCR Marker – 0.1 ml
SDP
Catalog No. 50811780
Encompass
The PCR Marker consists of a proprietary plasmid that is digested to completion with appropriate restriction enzymes to yield 5 double-stranded DNA bands suitable for use as molecular weight standards for agarose and acrylamide gel electrophoresis. The digested DNA includes fragments ranging from 50-766 base pairs. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Catalog No. 50-811-780 Supplier New England Biolabs, Inc. Supplier No. N3234S
The PCR Marker consists of a proprietary plasmid that is digested to completion with appropriate restriction enzymes to yield 5 double-stranded DNA bands suitable for use as molecular weight standards for agarose and acrylamide gel electrophoresis. The digested DNA includes fragments ranging from 50-766 base pairs. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.