Learn More
Biotang Inc PCR Master Mix. 2x50 rxns. Ready-to-use mixtures that include all of the reagents needed for PCR. Optimized for amplifying DNA template up to 3kb.Contains red and yellow loading dyes to allow loading of PCR product directly on a gel after thermal cyclin
Catalog No. 507513805 Shop All BIOTANG Inc Products
Encompass_Preferred
PCR Master Mix. 2x50 rxns. Ready-to-use mixtures that include all of the reagents needed for PCR. Optimized for amplifying DNA template up to 3kb.Contains red and yellow loading dyes to allow loading of PCR product directly on a gel after thermal cyclin
Catalog No. 50-751-3805 Supplier Biotang Inc Supplier No. P0311010
PCR Master Mix. 2x50 rxns. Ready-to-use mixtures that include all of the reagents needed for PCR. Optimized for amplifying DNA template up to 3kb.Contains red and yellow loading dyes to allow loading of PCR product directly on a gel after thermal cyclin