Learn More
Sigma Aldrich Fine Chemicals Biosciences PCR multiwell plates 96 we
Catalog No. 502900738
Encompass
A rigid top plate (included) minimizes plate distortion assures a dependable fit with the PCR thermal cycler and allows for a leak-proof seal with micro-mats or cap strips. Each well has a maximum capacity of 200 muL.
Catalog No. 50-290-0738 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3749031PAK
A rigid top plate (included) minimizes plate distortion assures a dependable fit with the PCR thermal cycler and allows for a leak-proof seal with micro-mats or cap strips. Each well has a maximum capacity of 200 muL.