Learn More
Apexbio Technology LLC PCR Mycoplasma Detection Kit(Synonyms: Nested PCR Mycoplasma Detection Kit, PCR Mycoplasma Test Kit, Mycoplasma PCR Detection Reagents), 100T.
SDP
Catalog No. 5000061109
Encompass_Preferred
PCR Mycoplasma Detection Kit employs nested PCR technology for identification of mycoplasma contamination in cell cultures Mycoplasmas represent small filterable prokaryotic microorganisms capable of profoundly affecting cellular gene and protein expression thus necessitating routine surveillance The kit targets the conserved ribosomal RNA operon regions within mycoplasma DNA initially amplifying a broad region (first PCR reaction) followed by amplification of a nested internal sequence (second PCR reaction) increasing detection specificity and sensitivity Researchers utilize this assay for regular monitoring and confirmation of mycoplasma contamination in culture systems
Catalog No. 50-000-61109 Supplier Apexbio Technology LLC Supplier No. K2821100
PCR Mycoplasma Detection Kit employs nested PCR technology for identification of mycoplasma contamination in cell cultures Mycoplasmas represent small filterable prokaryotic microorganisms capable of profoundly affecting cellular gene and protein expression thus necessitating routine surveillance The kit targets the conserved ribosomal RNA operon regions within mycoplasma DNA initially amplifying a broad region (first PCR reaction) followed by amplification of a nested internal sequence (second PCR reaction) increasing detection specificity and sensitivity Researchers utilize this assay for regular monitoring and confirmation of mycoplasma contamination in culture systems