Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ PCR Nucleotide Mix

Catalog No. BP25652 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP25652 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP25652

missing translation for 'validMainframeMsg'

A premixed 10mM solution of dATP, dCTP, dGTP, and dTTP used for PCR

  • Functionally tested in PCR
  • High purity: >98% triphosphate
  • Supplied as 10mM solution in purified water (pH 7.5)

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7.5
Content And Storage -20°C
Concentration 10 mM
Purity >98%
Quantity 200 μL
Color Colorless
For Use With (Application) PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.