Learn More
Sigma Aldrich Fine Chemicals Biosciences PCR Nucleotide Mix
Catalog No. 502906073
Encompass
PCR Nucleotide Mix is a clear colorless SOLUTIONon of the sodium salts of dATP dCTP dGTP and dTTP each at a concentration of 10 mM in Water PCR Grade. This nucleotide mixture can be added directly to polymerase chain reactions.
Catalog No. 50-290-6073 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11814362001
PCR Nucleotide Mix is a clear colorless SOLUTIONon of the sodium salts of dATP dCTP dGTP and dTTP each at a concentration of 10 mM in Water PCR Grade. This nucleotide mixture can be added directly to polymerase chain reactions.