Learn More
Sigma Aldrich Fine Chemicals Biosciences PCR Nucleotide Mix
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4638956001
Encompass
PCR Nucleotide Mix is a clear colorless SOLUTIONon of the sodium salts of dATP dCTP dGTP and dTTP each at a concentration of 10 mM in Water PCR Grade. This nucleotide mixture can be added directly to polymerase chain reactions.
Catalog No. 50-290-5165