Learn More
Sigma Aldrich Fine Chemicals Biosciences PCR Nucleotide MixPLUS
Catalog No. 502901132
Encompass
PCR Nucleotide MixPLUS is a clear colorless SOLUTIONon of the sodium salts of dATP dCTP dGTP each at a concentration of 10 mM and dUTP at a concentration of 30 mM in Water PCR Grade. This nucleotide mixture can be added directly to polymerase chain reactions.The incorporation of dUTP in place of dTTP allows the degradation of contaminating PCR products from former reactions with Uracil-DNA Glycosylase (UNG) to prevent carryover contamination from previous amplifications.
Catalog No. 50-290-1132 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11888412001
PCR Nucleotide MixPLUS is a clear colorless SOLUTIONon of the sodium salts of dATP dCTP dGTP each at a concentration of 10 mM and dUTP at a concentration of 30 mM in Water PCR Grade. This nucleotide mixture can be added directly to polymerase chain reactions.The incorporation of dUTP in place of dTTP allows the degradation of contaminating PCR products from former reactions with Uracil-DNA Glycosylase (UNG) to prevent carryover contamination from previous amplifications.