Learn More
GenDEPOT amfiSure ONE PCR Master Mix(2X), P7000, 5 x 1 mL
Catalog No. NC1561381
Encompass_Preferred
amfiSure ONE PCR Master Mix is ready-to-use mixtures that include all of the reagents needed for PCR. amfiSure PCR Master Mix provides superior yields in both routine and challenging PCR application such as direct PCR using crude extracted DNA, GC rich region PCR, colony PCR, multiplex PCR and etc.
Catalog No. NC1561381 Supplier GenDEPOT Supplier No. P7000005
amfiSure ONE PCR Master Mix is ready-to-use mixtures that include all of the reagents needed for PCR. amfiSure PCR Master Mix provides superior yields in both routine and challenging PCR application such as direct PCR using crude extracted DNA, GC rich region PCR, colony PCR, multiplex PCR and etc.