Learn More
Azzota Corp PCR plate 0.1ml 96W semi skirt
Catalog No. 5000886970
Encompass_Preferred
0 1ml 96 Well PCR Plate Semi Skirt Clear 25/pk 100pcs/Case
Catalog No. 50-008-86970 Supplier Azzota Corp Supplier No. PCRP01S100
Add to Cart
Add to Cart
0 1ml 96 Well PCR Plate Semi Skirt Clear 25/pk 100pcs/Case