Learn More
Mascon Medical PCR Plate, 96-well, 0.2 mL, Semi-Skirted, ABI, Clear, 10 pcs/pack, 5 packs/case
SDP
Catalog No. NC3527254
Encompass_Preferred
96-Well,flat top PCR microplate is designed to fitstandard 96 well blocks.Ultra-thin and consistentwall thickness allows precise thermal transfer foroptimum results.Can be sealed with 8/12 strip capsor sealing film.96-Well Polypropylene PCR Microplate, Compatible with ABI, Semi-skirted, Non-sterileFeaturesSuitable for 0.1 mL/0.2 mL thermal cycler blocksApplication:PCR,qPCR/Real-Time PCR
Catalog No. NC3527254 Supplier Mascon Medical Supplier No. MM802041
96-Well,flat top PCR microplate is designed to fitstandard 96 well blocks.Ultra-thin and consistentwall thickness allows precise thermal transfer foroptimum results.Can be sealed with 8/12 strip capsor sealing film.96-Well Polypropylene PCR Microplate, Compatible with ABI, Semi-skirted, Non-sterileFeaturesSuitable for 0.1 mL/0.2 mL thermal cycler blocksApplication:PCR,qPCR/Real-Time PCR