Learn More
USA Scientific Inc TempPlate non-skirted 24-well PCR plate, chimney, natural
Catalog No. NC9372279
Encompass_Preferred
TempPlate non-skirted polypropylene 0.2 mL 24-well PCR plate, chimney wells, standard depth, natural. 2/sleeve, 40/box.
Catalog No. NC9372279 Supplier USA Scientific Inc Supplier No. 14020240
Add to Cart
Add to Cart
TempPlate non-skirted polypropylene 0.2 mL 24-well PCR plate, chimney wells, standard depth, natural. 2/sleeve, 40/box.