Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PCR Plate, 384-well, standard, purple

Catalog No. FERAB1384P Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERAB1384P 50 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERAB1384P Supplier Thermo Scientific™ Supplier No. AB1384P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Standard Thermo Scientific™ 384-well plate, purple, is compatible with the majority of 384 block PCR and sequencing instruments.

The Standard Thermo Scientific™ 384-well plate, purple, is compatible with the majority of 384 block PCR and sequencing instruments. It has a maximum well volume of 40 μL and a working volume of 25 μL. This 384-well plate is compatible with all major 384 block thermal cyclers, and it has a cut corner at the A24 location.

Highlights of the Standard Thermo Scientific™ 384-well plate

  • Fully skirted for use with automated systems
  • Purple color to aid in sample tracking
  • Compatible with all leading 384 block thermal cyclers
  • 40 μL maximum well volume, 25 μL working volume
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut corner: A24

Applications

  • PCR
  • qPCR/Real-Time PCR

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Color Purple
Product Type PCR Microplate
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 40 μL
No. of Wells 384
Volume (Metric) Working 25 μL
Quantity 50 Plates
Reaction Speed Standard
Rimmed Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.