Learn More
Chemglass Life Sciences PCR Plate, Black Opaque, 96-Well, Skirted, 100µL
SDP
Catalog No. 501311314
Encompass_Preferred
PCR Plate, Black Opaque, 96-Well, Skirted, 100¿L. These opaque 96-well PCR plates are for chemiluminescent and fluorescent procedures. Each well has a capacity of 100¿L. Thin walled and designed for rapid thermal transfer. They are precision molded to ensure well-to-well and plate-to-plate uniformity.All sealing methods can be used for oil-free operation: domed and flat cap strips (CLS-3610 Series). Suitable for use with 96 well thermal cycler blocks that can accommodate full skirted plates, such as the MJ Research PTC 200.An alphanumeric grid helps in sample identification. To facilitate orientation, the top left corner of the plate is cut away. The AMPLATE is easy to seal since no cylindrical walls extend above the plate. Finally, the AMPLATE can be handled by robotic handling equipment and is ideal with automated pipetting systems. Autoclavable.10 plates packed in tamper-proof re-sealable bags, 100 per case.
Catalog No. 50-131-1314 Supplier Chemglass Life Sciences Supplier No. CLS361404B
Add to Cart
Add to Cart
PCR Plate, Black Opaque, 96-Well, Skirted, 100¿L. These opaque 96-well PCR plates are for chemiluminescent and fluorescent procedures. Each well has a capacity of 100¿L. Thin walled and designed for rapid thermal transfer. They are precision molded to ensure well-to-well and plate-to-plate uniformity.All sealing methods can be used for oil-free operation: domed and flat cap strips (CLS-3610 Series). Suitable for use with 96 well thermal cycler blocks that can accommodate full skirted plates, such as the MJ Research PTC 200.An alphanumeric grid helps in sample identification. To facilitate orientation, the top left corner of the plate is cut away. The AMPLATE is easy to seal since no cylindrical walls extend above the plate. Finally, the AMPLATE can be handled by robotic handling equipment and is ideal with automated pipetting systems. Autoclavable.10 plates packed in tamper-proof re-sealable bags, 100 per case.