Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PCR Plate, 96-well, semi-skirted, flat deck, blue

Catalog No. FERAB1400B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERAB1400B 25 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERAB1400B Supplier Thermo Scientific™ Supplier No. AB1400B
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Designed, manufactured, and tested to ensure PCR performance.

  • Compatible with most thermal cyclers and ABI platforms, including sequencers
  • No adapters necessary.
  • 0.3 mL maximum well volume when used with adhesive and heat seals.
  • Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
  • Blue color facilitates sample tracking
  • Flat plate deck facilitates sealing and handling
  • Available as SuperPlate providing 4X more rigidity for superior robotic handling
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 25 Plate
Color Blue
Product Type PCR Microplate
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Skirt Style Semi
Material Polypropylene
No. of Wells 96
Volume (Metric) Working 0.2 mL
Quantity 25 plates
Reaction Speed Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.