Learn More
CELLTREAT Scientific Products 96 Well PCR Plate, Half Skirt, Clear, 0.2mL
SDP
Catalog No. NC2131744
Encompass_Preferred
Boldly printed, alphanumeric markings for easy identification of individual wells. Universal design is compatible with the major brands of thermal cyclers. Thin-walled tubes provide uniform and efficient heat transfer. Certified RNase/DNase/DNA-free and non-pyrogenic to ensure reliable experimental results. Available in skirted and non-skirted options.
This item is not returnable. View return policy
Catalog No. NC2131744 Supplier CELLTREAT Scientific Products Supplier No. 229513
Boldly printed, alphanumeric markings for easy identification of individual wells. Universal design is compatible with the major brands of thermal cyclers. Thin-walled tubes provide uniform and efficient heat transfer. Certified RNase/DNase/DNA-free and non-pyrogenic to ensure reliable experimental results. Available in skirted and non-skirted options.