Learn More
Waters Corp PCR Plate Domino, 96-Position, Snug-Fitting, Machine-Readable
Supplier: Waters Corp 186010088
The 96-PCR Plate Domino is a machine-readable consumable holder designed to seamlessly integrate various PCR-type plates, both skirted and unskirted, with the Andrew+ pipetting robot. These dominos feature a snug-fitting 96x format, acting as a crucial interface for automating a wide range of laboratory workflows.
- Holds an array of PCR-type plates with or without skirts.
- Equipped with QR codes for critical labware information.
- Includes ARUCO codes for spatial positioning.
- Essential for any Andrew+ automation configuration.