Learn More
Research Products International Corp Simplate Low Profile PCR Plate, 96 Well, 140l Capacity, 50 per Case
SDP
Catalog No. 50476443
Encompass
Precision molded PCR plates ensure optimum uniformity and heat transfer. Thin, flexible design conforms to heating block wells and can be easily cut into smaller formats. Plates feature alphanumeric indexing, notched orientation corner and smooth top surface.
Plates accept various sealing materials such as films, micro-mats and caps. Natural color, autoclavable.
Catalog No. 50-476-443 Supplier Research Products International Corp Supplier No. 141328
Precision molded PCR plates ensure optimum uniformity and heat transfer. Thin, flexible design conforms to heating block wells and can be easily cut into smaller formats. Plates feature alphanumeric indexing, notched orientation corner and smooth top surface.
Plates accept various sealing materials such as films, micro-mats and caps. Natural color, autoclavable.