Learn More
Research Products International Corp PCR Plate Sealing Mat, 5 per Case
SDP
Catalog No. 50550166
Encompass
Ideal for sealing PCR plates used in thermal cyclers having ''pressure fit'' lids. Sealing Mat provides a secure, leak free seal during the PCR cycling process. Mat features identification tabs for proper alignment on plate and can be easily removed by lifting a corner tab. Made of clear silicone rubber for best chemical resistance including bleach solutions. Soft, pliable mat can be cut to fit 24, 32 or 48 format plates. Autoclavable. Packaged 5 per case.
Catalog No. 50-550-166 Supplier Research Products International Corp Supplier No. 141350
Ideal for sealing PCR plates used in thermal cyclers having ''pressure fit'' lids. Sealing Mat provides a secure, leak free seal during the PCR cycling process. Mat features identification tabs for proper alignment on plate and can be easily removed by lifting a corner tab. Made of clear silicone rubber for best chemical resistance including bleach solutions. Soft, pliable mat can be cut to fit 24, 32 or 48 format plates. Autoclavable. Packaged 5 per case.