Learn More
VWR INTERNATIONAL INC 96-Well PCR and Real Time PCR Plates, 200 uL, Natural, 96 wells, 10/pk
Catalog No. NC1503955
Encompass_Preferred
Polypropylene PCR plates are compatible with most thermal cyclers and are ideal for high-throughput screening thermocycler applications. Plates have a working capacity of 200 uL per well. They are thin, flexible, and easy to cut.
Catalog No. NC1503955 Supplier VWR INTERNATIONAL INC Supplier No. 82006704
Polypropylene PCR plates are compatible with most thermal cyclers and are ideal for high-throughput screening thermocycler applications. Plates have a working capacity of 200 uL per well. They are thin, flexible, and easy to cut.