Learn More
TWD Scientific LLC Pcr Plate, Twdbiox, 96 Well, Sterile, Polypropylene, Half Skirted, Flat Top Plate Edge, Printed Axis, 100/Case.
SDP
Catalog No. 502284993 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 96 Well Clear Polypropylene Non Sterile (Rnase & Dnase Free) Half Skirted Pcr Plate. Plates Come With Printed Axis, Flat Top Plate Edge, And Are Abi Compatible. Maximum Capacity Is >350µl. 10 Plates/Pack, 10 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-4993 Supplier TWD Scientific LLC Supplier No. BX603420
BioX 96 Well Clear Polypropylene Non Sterile (Rnase & Dnase Free) Half Skirted Pcr Plate. Plates Come With Printed Axis, Flat Top Plate Edge, And Are Abi Compatible. Maximum Capacity Is >350µl. 10 Plates/Pack, 10 Packs/Case, Price/Case