Learn More
TWD Scientific LLC Pcr Plate, Twdbiox, 96 Well, Sterile, Polypropylene, Half Skirted,Low Profile, 0.1Ml Raised Ridge, Printed Axis, 100/Case.
SDP
Catalog No. 502284995 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 96 Well Clear Polypropylene Non Sterile (Rnase & Dnase Free) Half Skirted Low Profile Pcr Plate For Fast. Plates Come With 0.1Ml Raised Rim, And Are Compatible With Abi 9800 & 7500. 10 Plates/Pack, 10 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-4995 Supplier TWD Scientific LLC Supplier No. BX603426
BioX 96 Well Clear Polypropylene Non Sterile (Rnase & Dnase Free) Half Skirted Low Profile Pcr Plate For Fast. Plates Come With 0.1Ml Raised Rim, And Are Compatible With Abi 9800 & 7500. 10 Plates/Pack, 10 Packs/Case, Price/Case