Learn More
TWD Scientific LLC Pcr Plate, Twdbiox, 96 Well, Polypropylene, Half Skited, Low Profile, Raised Edge And Rim, 100/Case.
SDP
Catalog No. 502284998 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 96 Well Clear Polypropylene Half Skirted Low Profile Pcr Plate For Lightcycler. Plates Come With Raised Plate Edge And Rim, Ultra Thin Walls, And Feature An H12-Position Cut Corner. 10 Plates/Pack, 10 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-4998 Supplier TWD Scientific LLC Supplier No. BX603455
BioX 96 Well Clear Polypropylene Half Skirted Low Profile Pcr Plate For Lightcycler. Plates Come With Raised Plate Edge And Rim, Ultra Thin Walls, And Feature An H12-Position Cut Corner. 10 Plates/Pack, 10 Packs/Case, Price/Case