Learn More
Biotang Inc 96-wellPCR plate, Skirt Natural.100/case. For use with Eppendorf, Biorad, MJ thermal cyclers and DNA analyzers. Skirt allows robotic handling and sample labeling.Real-time PCR compatible. Dnase, Rnase, Human DNA, Pyrogen, PCR Inhibitors Free
Catalog No. 507515019 Shop All BIOTANG Inc Products
Encompass_Preferred
96-wellPCR plate, Skirt Natural.100/case. For use with Eppendorf, Biorad, MJ thermal cyclers and DNA analyzers. Skirt allows robotic handling and sample labeling.Real-time PCR compatible. Dnase, Rnase, Human DNA, Pyrogen, PCR Inhibitors Free
Catalog No. 50-751-5019 Supplier Biotang Inc Supplier No. BT28440
96-wellPCR plate, Skirt Natural.100/case. For use with Eppendorf, Biorad, MJ thermal cyclers and DNA analyzers. Skirt allows robotic handling and sample labeling.Real-time PCR compatible. Dnase, Rnase, Human DNA, Pyrogen, PCR Inhibitors Free