Learn More
Cole-Parmer PCR preparation rack, assorted colors, 5/pk
Supplier:  Cole-Parmer 0673607
Encompass
PCR preparation rack, Polypropylene, assorted colors, 5/pk, Compact polypropylene (PP) racks are designed for preparing and storing 0.2 ml PCR strips and tubes for sample processing. Arranged in an 8 x 12 alphanumeric grid the 96 wells keep tubes upright, secure and provide for easy sample identification. Translucent PP covers allow the samples to be viewed without removal. Autoclavable at 250°F (121°C).
Catalog No. NC1046244